Handheld digital microscopes, Dino-Lite High Speed (VGA / D-SUB)
Proveedor: Dino-Lite / IDCP B.V.
AM5116T AM5116ZT AM5116ZTL AM5216ZT AM5216ZTL AM5216TF
630-1857EA 479 EUR
630-1857 630-1861 630-1862 IDCPAM5216ZT IDCPAM5216ZTL IDCPAM5216TF
Handheld digital microscopes, Dino-Lite High Speed (VGA / D-SUB)
Microscopios
High Speed Real Time models connect directly to a computer monitor/ VGA screen via a VGA (D-SUB) connector. Ideal for use in situations where real time images are vital.
  • High frame rate of up to 60 fps at 800×600 resolution
  • Composite/plastic housing; models AM4016MZT and AM4016MZTL feature a metal housing and are therefore ESD-safe
  • Edge series models: High-quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
  • Integrated polariser greatly reduces glare on reflective surfaces (some models only)
  • Model AM52