Anti-Glial Fibrillary Acidic Protein (GFAP) (G-A-5) Mouse Monoclonal Antibody

Proveedor: Merck Millipore (Oncogene)
IF03L-100
IF03L-100EA 589 EUR
IF03L-100
Anti-Glial Fibrillary Acidic Protein (GFAP) (G-A-5) Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: Glial Fibrillary Acidic Protein (GFAP) (G-A-5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

ESPECIFICACIONES

Simbolo antígeno GFAP
Environmentally Preferable
Nombre del antígeno Glial fibrillary acidic protein
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Reactividad
Huésped Mouse
Isotipo
Western blot Yes
InmunoQuímica Yes
Precipitación inmunológica Yes
FluorescenciaInmunológica Yes
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR