Magnetic hotplate stirrer RCT 5 digital
Proveedor: IKA
20002704
IKAA20002704EA 1270 EUR
IKAA20002704
Magnetic hotplate stirrer RCT 5 digital
Imánes de agitación Agitadores magnéticos con placas calefactoras
The RCT 5 digital magnetic hotplate stirrer offers significantly higher performance with heat output of approximately 800 W. This hotplate stirrer is ideal for a wide range of heating and stirring tasks.
  • Automatic reverse rotation with intermittend mode
  • Multifunctional menu
  • Viscosity trend measurement
  • Digital LED display shows actual and set speed setting
  • Timer and sensor in medium detection (via PT1000 sensor)
  • Can be used at 5 to 40 °C (80% relative humidity), in an incubator or cold room
Whether determining the direction of rotation, timer functionsMore Product Information