Temperature control systems
Proveedor: IKA
8035900 8035800 8036000 8036100 8036200 8036300 8036400 8035700 8036800 8037200 10000416 10000415 10002471 10002474
IKAA8035900EA 3200 EUR
IKAA8035900 IKAA8035800 IKAA8036000 IKAA8036100 IKAA8036200 IKAA8036300 IKAA8036400 IKAA8035700 IKAA8036800 IKAA8037200 IKAA10000416 IKAA10000415 IKAA10002471 IKAA10002474
Temperature control systems
Termostatos con circulación Sistemas de circulación cerrada Sistemas de control de temperatura
ICC compact immersion circulators allow for flexible coupling to water baths of different volumes. They are designed for tempering liquids up to temperatures of 150 °C. The convenient carrying handle and compact design allow easy transportation as well as comfortable use. An economic and attractive solution for basic applications such as sample tempering and glassware heating.
  • Adjustable safety circuit
  • Switch from internal to external temperature with one key (only for control unit)
  • Optical and acoustic warning, example: For overheating
  • Powerful pressure/