Homogeniser, T 18 digital ULTRA-TURRAX®
Proveedor: IKA
3720003 3720000 3720002 9022802 9022800
431-0187EA 1180 EUR
431-0187 431-0183 431-0186 431-0190 IKAA9022800
Homogeniser, T 18 digital ULTRA-TURRAX®
Homogenizadores Homogeneizadores de sobremesa
Stand model, volume range 1 to 1500 ml. The T 18 digital drive unit offers a wide speed range from 3000 to 25000 min⁻¹ that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantees a wide range of applications.
  • Digital speed display
  • Electronic speed control and overload protection
  • Stainless steel dispersing elements can be cleaned quickly and easily
  • Plastic disposable dispersing elements are available in two sizes
  • Error code display
IP protection class according to DIN EN 605More Product Information