Manual Vacuum Regulation Turret, Flush Mount
Proveedor: Welch by Gardner Denver
700562
ILMV700562EA 267 EUR
ILMV700562
Manual Vacuum Regulation Turret, Flush Mount
Válvulas Válvulas de vacío
Open/close the vacuum line and easily control flow rate.
  • Dimensions: 690 mm
  • Diameter: 820 mm protrusion from wall and 20 mm into wall
Turret has stepped hose barb that accepts 6,35 - 9,52 mm vacuum hose (DN 8-10).