Anti-MMP-1 (Ab-1) (41-1E5) Mouse Monoclonal Antibody

Proveedor: Merck Millipore (Oncogene)
IM35-100
IM35-100EA 589 EUR
IM35-100
Anti-MMP-1 (Ab-1) (41-1E5) Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: MMP-1 (Ab-1) (41-1E5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno MMP-1
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Reactividad Human
Huésped Mouse
Isotipo
Western blot Yes
InmunoQuímica Yes
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR