Anti-MMP-2 (Ab-4) (75-7F7) Mouse Monoclonal Antibody [clone: 75-7F7]

Proveedor: Merck Millipore (Oncogene)
IM51-100
IM51-100EA 577 EUR
IM51-100
Anti-MMP-2 (Ab-4) (75-7F7) Mouse Monoclonal Antibody [clone: 75-7F7]
Anticuerpos
Type: Primary
Antigen: MMP-2 (Ab-4) (75-7F7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR