tempmate.®-S1 Single Use Temperature Data Logger, L1 2 °C, H1 8 °C, SD 30 mins
Proveedor: tempmate
S1C15A69010
IMECS1C15A69010EA 461 EUR
IMECS1C15A69010
tempmate.®-S1 Single Use Temperature Data Logger, L1 2 °C, H1 8 °C, SD 30 mins
Registradores de datos
Pre-programmed. Outstanding single-use temperature logger for reliable cold chain monitoring. Integrated USB port and PDF and CSV generating software on-board.
  • Pre-programmed
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food grade packing
  • No additional hardware or software required
  • Detailed PDF and CSV report including graph and table
  • Intelligent LEDs for status monitoring
  • Extended shelf-life of two years
tempmate.®-S1 loggers are specially designed for temperature monitoring in any cold chain logistics. As a pre-More Product Information