Anti-IgG Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))
Proveedor: ImmunoReagents
IR1126
IMMRIR1126EA 186 EUR
IMMRIR1126
Anti-IgG Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
ImmunoReagents’ Alkaline Phosphatase (AP) antibody conjugates are used in ELISA, Immunohistochemistry (IHC) and Western blotting techniques. AP is a 140 kDa enzyme that changes color in the presence of PNPP or BCIP/NBT substrates. These conjugates have a linear reaction rate thus allowing the researcher to increase sensitivity by prolonging substrate incubation times.
  • Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will beMore Product Information