Anti-IgG Donkey Polyclonal Antibody (DyLight® 633)
Proveedor: ImmunoReagents
IR1176
IMMRIR1176EA 423 EUR
IMMRIR1176
Anti-IgG Donkey Polyclonal Antibody (DyLight® 633)
Anticuerpos
The ImmunoReagents' DyLight® 633 conjugates fluoresces red with physical properties comparable to other 633 dyes. DyLight® 633 is ideal for multi-color applications. DyLight® 633 is available as reactive labeling agents and as conjugates of secondary antibodies and biotin-binding proteins for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 638 nm / 658 nm Emission Color = Red (Similar Dyes: Alexa Fluor 633)
  • Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formatsMore Product Information