Anti-IgG Donkey Polyclonal Antibody (DyLight® 594)
Proveedor: ImmunoReagents
IR1313
IMMRIR1313EA 135 EUR
IMMRIR1313
Anti-IgG Donkey Polyclonal Antibody (DyLight® 594)
Anticuerpos
The ImmunoReagents' DyLight® 594 conjugates generate red fluorescence and shows wide excitation wavelength separation from green fluorophores for multi-Colour applications with reduced bleedthrough. The physical characteristics of DyLight® 594 are comparable to other 594 dyes. DyLight® 594 is available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 593 nm/618 nm Emission Colour = Red (Similar Dyes: Alexa Fluor 594, Texas Red).
  • Origin: US
ImmunoReagents' antibodies are suitable forMore Product Information