Anti-kappa light chain Goat Polyclonal Antibody (DyLight® 650)
Proveedor: ImmunoReagents
IR1500
IMMRIR1500EA 400 EUR
IMMRIR1500
Anti-kappa light chain Goat Polyclonal Antibody (DyLight® 650)
Anticuerpos
The ImmunoReagents' DyLight® 650 conjugates provide vibrant far-red fluorescence with comparable or improved performance compared to other dyes for fluorescent applications. DyLight® 650 dye conjugates have tight excitation/emission spectra that are ideal for multi-Colour applications. DyLight® 650 conjugated secondary antibodies are available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 652 nm/672 nm Emission Colour = Far Red (Similar Dyes: Alexa Fluor 647, Cy5).
  • Origin: US