Anti-kappa light chain Goat Polyclonal Antibody (DyLight® 800)

Proveedor: ImmunoReagents
IR1501
IMMRIR1501EA 322 EUR
IMMRIR1501
Anti-kappa light chain Goat Polyclonal Antibody (DyLight® 800)
Anticuerpos
The ImmunoReagents' DyLight® 800 conjugates uses a near-IR fluorphore that is invisible to the naked eye but increases the staining options when using infrared imaging systems. DyLight® 800 has spectral properties that are very similar to other near-IR dyes, including Alexa Fluor 790 and IRDye 800, and has high solubility for maximal incorporation of the dye into the biomolecule of interest. Excitation/Emission = 777 nm/794 nm Emission Colour = Near IR (Similar Dyes: Alexa Fluor 790, IRDye 800).

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats andMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno Ig kappa
Nombre del antígeno Kappa Light Chain
Environmentally Preferable
Tipo de anticuerpo Secundario
Clonalidad Polyclonal
Conjugado DyLight® 800
Reactividad Human
Huésped Goat
Isotipo IgG
Western blot Yes
ELISA Yes
InmunoQuímica Yes
Precipitación inmunológica Yes
FluorescenciaInmunológica Yes
Citometría de flujo Yes
Epítopo Light Chain
Inmunógeno Human kappa (k) light chain
Formato Purified IgG
Forma Lyophilised
Purification Affinity purified using solid phase Human Kappa (к) Chain
Concentración 1,0 mg/ml (E 1% at 280 nm = 13,0)
Adsorción cruzada No
Storage buffer 10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1% (w/v) BSA, Protease/IgG free
Temperatura de almacenamiento 2...8 °C
Shipping temperature 2...8 °C
Tested applications Fluoresence ELISA, Fluorescence-Activated Cell Sorting

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR