Anti-IgA alpha Goat Polyclonal Antibody (Biotin (NHS-LC))

Proveedor: ImmunoReagents
IR1515
IMMRIR1515EA 243 EUR
IMMRIR1515
Anti-IgA alpha Goat Polyclonal Antibody (Biotin (NHS-LC))
Anticuerpos
ImmunoReagents Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualized, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.

  • Origin: US

ImmunoReagents antibodies are suitableMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno IgA
Nombre del antígeno Immunoglobulin A alpha
Environmentally Preferable
Tipo de anticuerpo Secundario
Clonalidad Polyclonal
Conjugado Biotin (NHS-LC)
Reactividad Human
Huésped Goat
Isotipo IgA
Western blot Yes
ELISA Yes
InmunoQuímica Yes
Precipitación inmunológica Yes
FluorescenciaInmunológica Yes
Citometría de flujo Yes
Fragmento de anticuerpo F(ab')2
Inmunógeno Human IgA, (α chain)
Formato Purified IgG
Forma Clear, colorless liquid
Purification Affinity purified using solid phase Human IgA
Concentración 1,50 mg/ml (E 1% at 280 nm = 13,0)
Adsorción cruzada No
Storage buffer 10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
Temperatura de almacenamiento 2…8 °C
Shipping temperature 2…8 °C
Tested applications Fluorescence-Activated Cell Sorting

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR