Anti-IgG Goat Polyclonal Antibody

Proveedor: ImmunoReagents
IR2926
IMMRIR2926EA 83.4 EUR
IMMRIR2926
Anti-IgG Goat Polyclonal Antibody
Anticuerpos
  

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR