Anti-IgG Rabbit Polyclonal Antibody (Biotin)

Proveedor: ImmunoReagents
RT-003-ZBIO
IMMRRT-003-ZBIOEA 103 EUR
IMMRRT-003-ZBIO
Anti-IgG Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR