Azul nitro-tetrazolio cloruro (NBT) ≥99%

Proveedor: Thermo Fisher Scientific
Atención

J60230.03 J60230.06
J60230.03EA 111 EUR
J60230.03 J60230.06
Azul nitro-tetrazolio cloruro (NBT) ≥99%
Azul nitro-tetrazolio cloruro (NBT)

Purity: 99% CAS No.: 298-83-9 Molecular Formula: C40H30Cl2N10O6 Formula Weight: 817.65 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Melting Point: ca 190° dec. Beilstein Reference: 4115923 MDL No.: MFCD00012159


Intense blue dye used in histochemistry determinations

Formula: C₄₀H₃₀Cl₂N₁₀O₆
Pto. de fusión: ∼ 190 °C (desc.)
Temperatura de almacenaje: Nevera
Número MDL: MFCD00012159
Núm. CAS: 298-83-9
EINECS: 206-067-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR