Kanamicina sulfato cell culture reagent

Proveedor: Thermo Fisher Scientific
Peligro

J60668.03 J60668.06
J60668.03EA 62.4 EUR
J60668.03 J60668.06
Kanamicina sulfato cell culture reagent
Kanamicin sulfato

CAS No.: 25389-94-0 Molecular Formula: C18H36N4O11.H2SO4 Formula Weight: 582.58 Physical Form: Powder Appearance: White to off-white Solubility: Soluble in water at 10mg/ml MDL No.: MFCD00070253

Formula: C₁₈H₃₈N₄O₁₅S
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00070253
Núm. CAS: 25389-94-0
EINECS: 246-933-9
Índice Merck: 14,5281

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR