Rifampicina para biología molecular

Proveedor: Thermo Fisher Scientific
Atención

Sinónimos: Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP

J60836.03 J60836.06 J60836.14
J60836.03EA 54.1 EUR
J60836.03 J60836.06 J60836.14
Rifampicina para biología molecular
Rifampicina

CAS No.: 13292-46-1 Molecular Formula: C43H58N4O12 Formula Weight: 822.94 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Faint orange to red/brown Melting Point: 183-188° Solubility: Soluble in DMSO or methanol Beilstein Reference: 5723476 Merck Reference: 14,8216 MDL No.: MFCD00151389


Antibiotic effective against mycobacteria. Can also inhibit DNA polymerase

Formula: C₄₃H₅₈N₄O₁₂
MW: 822.95 g/mol
Pto. de fusión: 183…188 °C
Temperatura de almacenaje: Congelador
Número MDL: MFCD00151389
Núm. CAS: 13292-46-1
EINECS: 236-312-0
Índice Merck: 14,8216

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR