Miconazole 97%

Proveedor: Thermo Fisher Scientific
Atención

J60872.03 J60872.06 J60872.14
J60872.03EA 37.7 EUR
J60872.03 J60872.06 J60872.14
Miconazole 97%
Miconazole

Purity: 99% CAS No.: 22916-47-8 Molecular Formula: C18H14Cl4N2O Formula Weight: 416.13 Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to off-white Melting Point: 170° Solubility: Freely soluble in alcohols or acetone. Also soluble in DMF or chloroform. Insoluble in water MDL No.: MFCD00216019


Applications: Inhibits cytochrome P450-dependent 14-alpha-demethylase

Formula: C₁₈H₁₄Cl₄N₂O
Pto. de fusión: 83…97 °C
Número MDL: MFCD00216019
Núm. CAS: 22916-47-8
EINECS: 245-324-5
Índice Merck: 14,6178

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR