Ácido 5,8,11,14-Eicosatetrainóico

Proveedor: Thermo Fisher Scientific

J61624.MC J61624.FX
J61624.MCEA 631 EUR
J61624.MC J61624.FX
Ácido 5,8,11,14-Eicosatetrainóico
Ácido 5,8,11,14-Eicosatetrainóico

CAS No.: 1191-85-1 Molecular Formula: C20H24O2 Formula Weight: 296.40 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Off-white Melting Point: 81-82° Beilstein Reference: 1798411 MDL No.: MFCD00036967


A cyclooxygenase, lipoxygenase and cytochrome P450 inhibitor

Formula: C₂₀H₂₄O₂
Pto. de fusión: 81…82 °C
Número MDL: MFCD00036967
Núm. CAS: 1191-85-1

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR