Papain (from Carica Papaya Latex)
Proveedor: Thermo Fisher Scientific
J61875.MB J61875.MC J61875.03
J61875.MBEA 88.8 EUR
J61875.MB J61875.MC J61875.03
Papain (from Carica Papaya Latex)
Enzimas
Applications: A cysteine protease; breaks down intercellular matrix of cartilage