L-Cystine dihydrochloride 99%

Proveedor: Thermo Fisher Scientific

J62292.14 J62292.22 J62292.30
J62292.14EA 77.5 EUR
J62292.14 J62292.22 J62292.30
L-Cystine dihydrochloride 99%
L-Cystine dihydrochloride

Purity: 99% CAS No.: 30925-07-6 Molecular Formula: C6H12N2O4S2.2HCl Formula Weight: 313.22 Physical Form: Powder Appearance: White to pale yellow Melting Point: 228-232° MDL No.: MFCD00070399

Formula: C₆H₁₂N₂O₄S₂·2HCl
Pto. de fusión: 228…232 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00070399
Núm. CAS: 30925-07-6
EINECS: 250-391-9

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR