L(+)-Leucina cell culture reagent

Proveedor: Thermo Fisher Scientific

J62824.22 J62824.30 J62824.36
J62824.22EA 93.6 EUR
J62824.22 J62824.30 J62824.36
L(+)-Leucina cell culture reagent
L(+)-Leucina

CAS No.: 61-90-5 Molecular Formula: C6H13NO2 Formula Weight: 131.17 Physical Form: Powder Density: 1.293 Appearance: White to off-white Melting Point: 300° Beilstein Reference: 1721722 Merck Reference: 14,5451 MDL No.: MFCD00002617

Formula: C₆H₁₃NO₂
Pto. de fusión: >300 °C
Densidad: 1,293
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00002617
Núm. CAS: 61-90-5
EINECS: 200-522-0
Índice Merck: 14,5451

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR