α-Ketoglutaric acid disodium salt dihydrate ≥99%

Proveedor: Thermo Fisher Scientific

Sinónimos: 2-Oxoglutaric acid disodium salt dihydrate, α-Ketoglutaric acid disodium salt dihydrate, Sodium α-ketoglutarate dibasic dihydrate

J63154.03 J63154.14 J63154.22 J63154.30
J63154.03EA 21.2 EUR
J63154.03 J63154.14 J63154.22 J63154.30
α-Ketoglutaric acid disodium salt dihydrate ≥99%
α-Ketoglutaric acid disodium salt dihydrate

Purity: 99% CAS No.: 305-72-6 Molecular Formula: C5H4Na2O5.2H2O Formula Weight: 226.09 (190.06anhy) Physical Form: Powder Sensitivity: Hygroscopic Appearance: White MDL No.: MFCD00150702

Formula: C₅H₄Na₂O₅·2H₂O
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00150702
Núm. CAS: 305-72-6
EINECS: 206-167-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR