Citrate-buffered saline (20X)

Proveedor: Thermo Fisher Scientific

J63666.K2 J63666.K3
J63666.K2EA 36.9 EUR
J63666.K2 J63666.K3
Citrate-buffered saline (20X)
Tampones Tampones para electroforesis Tampones de hibridación

Saline-sodium citrate (SSC) buffer.


300 mM sodium citrate, 3M sodium chloride, pH 7.0

Order Now

ESPECIFICACIONES

Environmentally Preferable
Physical form Liquid
MDL No. MFCD00132371

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR