Buffers, Sodium Citrate, 0,5M Solutions

Proveedor: Thermo Fisher Scientific

J62918.AK J62918.AP J61815.AK J61815.AP J63888.AK J63888.AP J63199.AK J63199.AP
J62918.AKEA 80 EUR
J62918.AK J62918.AP J61815.AK J61815.AP J63888.AK J63888.AP J63199.AK J63199.AP
Buffers, Sodium Citrate, 0,5M Solutions
Tampones Tampones biológicos

Storage temperature: 2 to 8 °C
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00012462

Order Now

ESPECIFICACIONES

Environmentally Preferable
Odour/scent Odourless
Physical form Liquid
MDL No. MFCD00012462
Apariencia Colourless
Temperatura de almacenamiento 2°C to 8°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR