Lactobionic acid 97% (by titrimetric analysis)

Proveedor: Thermo Fisher Scientific

J66368.14 J66368.22 J66368.30
J66368.14EA 97.3 EUR
J66368.14 J66368.22 J66368.30
Lactobionic acid 97% (by titrimetric analysis)
Lactobionic acid

Soluble in water

Formula: C₁₂H₂₂O₁₂
Pto. de fusión: 113…118 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00078147
Núm. CAS: 96-82-2
EINECS: 202-538-3
Índice Merck: 14,5342

Order Now

Resultados test especificaciones

Colour White to Off-White
Form Powder
Assay (Titration) 97% min
Water Content (Karl Fischer Titration) 5,0% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR