Oro 99% supplied in 0.1 mM PBS, nanopartículas 250 nm reactant-free

Proveedor: Thermo Fisher Scientific

J67457.ACEA 169 EUR
J67457.AC
Oro 99% supplied in 0.1 mM PBS, nanopartículas 250 nm reactant-free
Oro

Purity: 99% CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Dark brown to pink MDL No.: MFCD00003436

Formula: Au
Temperatura de almacenaje: Nevera
Número MDL: MFCD00003436
Núm. CAS: 7440-57-5

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR