Linear polyacrylamide solución 5 mg/ml para biología molecular, for DNA/RNA precipitation
Proveedor: Thermo Fisher Scientific
Sinónimos: LPA solution for DNA/RNA precipitation
J67830.XF
J67830.XFEA 101 EUR
J67830.XF
Linear polyacrylamide solución 5 mg/ml para biología molecular, for DNA/RNA precipitation
Linear polyacrylamide
Linear polyacrylamide solution for DNA/RNA precipitation, 5mg/ml, Molecular Biology Grade. Liquid.
Storage Temperature: -30°C to -10°C Physical Form: Liquid Appearance: Clear, Colourless