Accesorios para contenedores de residuos de riesgo biológico
Proveedor: JUSTRITE MANUFACTURING
05901
JUSU05901EA 112 EUR
JUSU05901
Accesorios para contenedores de residuos de riesgo biológico
Bolsas Bolsas de eliminación de residuos
For safe, temporary collection of biological waste or contaminated materials. Red color is easily identifiable as biohazard bags.
  • 15 Gallon (56,8 L) capacity bag fits Justrite Biohazard cans of 6 and 10 Gallon sizes
  • High density PE of 11 microns thickness provides secure handling to avoid punctures and tears
  • Tubular construction with a single side seal and star seal bottom provide a stable, leak proof bag
  • Specifically labeled for Biohazard material handling
Printing includes English, Spanish and French translation along with the international recognizable biohazard symbol.