Drum Equipment
Proveedor: JUSTRITE MANUFACTURING
08916
JUSU08916EA 256 EUR
JUSU08916
Drum Equipment
Almacén
Safety technology for drum storage and dispensing.
Proper drum storage and dispensing protect personnel and property from fire hazards.
Certificaciones: Many items meet OSHA, FM and TUV regulations.