Maxima™ Probe qPCR Master Mix (2X)
Proveedor: Thermo Fisher Scientific
K0231 K0232 K0233 K0261 K0262 K0263
K0231EA 239 EUR
K0231 K0232 K0233 K0261 K0262 K0263
Maxima™ Probe qPCR Master Mix (2X)
Reactivos de ácidos nucleicos qPCR/RT-qPCR Enzymes and Kits
The Maxima™ Probe qPCR Master Mix (2X) is a ready to use solution optimised for quantitative real time PCR and two-step real time RT-PCR on most real time PCR machines. The master mix includes Maxima™ hot start Taq DNA polymerase and dNTPs in an optimised PCR buffer. dUTP is included in the mix for optional carryover contamination control using uracil-DNA glycosylase (UDG). The use of Maxima™ probe qPCR master mix (2X) in real time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates.
  • Specificity – Maxima™ hot start Taq DNA polymerase in combination with the optimised buffer eliminates non-specific amplification and formation of primer dimers
  • Sensitivity – detects low copy number targets
  • Wide linear range – accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience – ready to use
Información de suministro: Maxima Probe/ROX qPCR Master Mix (2X) is supplemented with ROX passive reference dye. Maxima Probe qPCR Master Mix, ROX solution provided (2X), comes with a separate ROX vial. Nuclease-free water is also provided.
Order Now
ESPECIFICACIONES
Environmentally Preferable
Aplicación sonda qPCR