Super-Duty SMS/Film Steam Sterilisation Tubings
Proveedor: KEYSTONE CLEANROOM PRODUCTS
TUBRPKC600-12 TUBRPKC600-14 TUBRPKC600-18 TUBRPKC600-24 TUBRPKC600-28 TUBRPKC600-4 TUBRPKC600-6 TUBRPKC600-9
KACCTUBRPKC600-12EA 1330 EUR
KACCTUBRPKC600-12 KACCTUBRPKC600-14 KACCTUBRPKC600-18 KACCTUBRPKC600-24 KACCTUBRPKC600-28 KACCTUBRPKC600-4 KACCTUBRPKC600-6 KACCTUBRPKC600-9
Super-Duty SMS/Film Steam Sterilisation Tubings
Bolsas Bolsas de esterilización
Ideal for applications where the use of standard SMS wraps prevents the user from being able to see and identify the contents of the package.
  • 88 gram SMS weight
  • The tubing form allows for sizing flexibility
  • The ends of the tubing should be heat sealed closed
  • When opened, virtually no particulates are generated
  • Manufactured in a certified ISO6 cleanroom
Made from spunbond/meltblown/spunbond polypropylene fabric and a polyester-based flexible film. The SMS side is an effective bacterial barrier, post sterilisation. The clear film side offers excellentMore Product Information