Overturnable Stoppers, Silicone Rubber
Proveedor: KARTELL
0119100 0119300 0119400 0119500 0119600 0119700 0119000 0119200
KART0119100EA 64.4 EUR
KART0119100 KART0119300 KART0119400 KART0119500 KART0119600 KART0119700 KART0119000 KART0119200
Overturnable Stoppers, Silicone Rubber
Tapones Tapones de Silicona
Pierceable stoppers to withdraw solutions and suspensions from closed containers, using syringes.
Suitable for use in food and beverage industry.
Height (H/h): H: height of overturnable section; h: height of lower section.