Tubos, PVC, K30
Proveedor: KARTELL
3890 3891 3892 3893 3894 3895 3896
KART3890EA 25.4 EUR
KART3890 KART3891 KART3892 KART3893 KART3894 KART3895 KART3896
Tubos, PVC, K30
Tuberías Tubo de transferencia
PVC, transparent. Flexible and transparent hoses manufactured in atoxic material.
  • Suitable for liquids transfer
  • Cadmium free
  • Temperature resistance: -5 to +50 °C
Shore hardness: 65 ±5