Douche cans/irrigators
Proveedor: KARTELL
590
KART590EA 10.4 EUR
KART590
Douche cans/irrigators
Contenedores
Douche cans/irrigators, made of polypropylene.
  • Translucent
  • With flat back and suspension tag
  • Autoclavable