Magnetic stirring bars, PTFE, oval
Proveedor: KARTELL
683 684 685 686 687 688 689
KART683EA 9.1 EUR
KART683 KART684 KART685 KART686 KART687 KART688 KART689
Magnetic stirring bars, PTFE, oval
Imánes de agitación Barras magnéticas de agitación
These oval shaped PTFE stirring bars are ideal for containers or bottles with round bottom.
  • Suitable for food stuff