Bulb pipettes, with safety bulb, Qualicolor, colour-coded, one mark, Class AS
Proveedor: Kavalier
632433311025
KAVA632433311025EA 26.3 EUR
KAVA632433311025
Bulb pipettes, with safety bulb, Qualicolor, colour-coded, one mark, Class AS
Pipetas Pipetas aforadas
Soda-lime glass.
  • Enlarged form
  • Not autoclavable at 121 °C
Certificaciones: DIN 12691, ISO 648