Heat and cold resistant gloves with chemical protection, Tricotril® Winter
Proveedor: KCL by Honeywell
739.8 739.10 739.11 739.9
KCHL739.8EA 1360 EUR
KCHL739.8 KCHL739.10 KCHL739.11 KCHL739.9
Heat and cold resistant gloves with chemical protection, Tricotril® Winter
Guantes
These gloves are resistant to a variety of chemicals and provide short-term protection against cold down to −20 °C and against heat up to 250 °C. The heavy seamless wool liners are fully nitrile coated.
  • Broad chemical resistance, high mechanical resistance
  • Textured profile for good grip when handling greasy and oily parts
  • Free from natural latex
  • Good antistatic and electrostatic properties
  • Long cuff also protects forearm
Ideal applications include working in cold areas and environments with steam hazards, handling of contaminated heavy parts, cleaningMore Product Information