Blood collection tubes with lithium heparin
Proveedor: KIMA SAS DI CHIARIN RENZO
18648
KIMA18648EA 0 EUR
KIMA18648
Blood collection tubes with lithium heparin
Sistemas para diagnóstico Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Polypropylene test tube with stopper.
  • Lithium heparin for 5 ml of blood
  • Non sterile
  • With writing area