Dialysis tubing Spectra/Por®, biotech CE
Proveedor: KLEINFELD
131450 131450T
KLEI131450EA 1060 EUR
KLEI131450 KLEI131450T
Dialysis tubing Spectra/Por®, biotech CE
Tuberías Tuberías para diálisis
These cellulose Ester (CE) membrane offers the largest range of concise for desalting, isolating ionic species and macromolecular purifications which is supplied wet in 0,05 % or 0,1% sodium azide.
  • Superior chemical compatibility and temperature tolerance
  • Rigidly controlled porosity for better selectivity and separations
  • Ultrapure with no trace heavy metals & sulfides
  • Does not interfere with molecular activity and function
  • No glycerin removal or pretreatment required
Tubing is more sensitive to harsh conditions and solvents and generally toleratesMore Product Information