Dess-Martin periodinane

Proveedor: Thermo Fisher Scientific
Peligro

Sinónimos: 1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one, DMP, (1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl ester

L15779.03 L15779.06 L15779.14
L15779.03EA 49.3 EUR
L15779.03 L15779.06 L15779.14
Dess-Martin periodinane
Dess-Martin periodinane

Dess Martin periodinane is used to oxidize primary alcohols to aldehydes and secondary alcohols to ketones. It is also used as an oxidant for complexes with multifunctional alcohols. It is actively involved in the oxidation of N-protected-amino alcohols without epimerization and allylic alcohols.


CAS No.: 87413-09-0 Molecular Formula: C13H13IO8 Formula Weight: 424.15 Storage Temperature: -30°C to -10°C Melting Point: 130-132° dec. Beilstein Reference: 4548207 UN No.: UN1479 MDL No.: MFCD00130127

Formula: C₁₃H₁₃IO₈
MW: 424.15 g/mol
Pto. de fusión: 130…132 °C (desc.)
Número MDL: MFCD00130127
Núm. CAS: 87413-09-0
ADR: 5.1 III

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR