Drums, narrow neck
Proveedor: LAMAPLAST
207
LAMA207EA 50.2 EUR
LAMA207
Drums, narrow neck
Barriles, Barriles, Tambores
HDPE, narrow neck
  • Good chemical resistance
  • Good temperature resistance
  • Stackable
  • Self sealing caps with total polyurethrane foam gasket
Ideal for use with foods.