Jerricans, narrow neck, HDPE, with screwed stopcock
Proveedor: LAMAPLAST
3963 3961S
LAMA3963EA 36.3 EUR
LAMA3963 LAMA3961S
Jerricans, narrow neck, HDPE, with screwed stopcock
Frascos Bidones
HDPE, natural.
  • Supplied with self sealing plastirob stopcock
  • Self sealing cap
  • Good chemical and temperature resistance
Ideal for use in food industries.