Calibration thermostats, ECO
Proveedor: LAUDA
L002847 L002848 L002850
LAUDL002847EA 10900 EUR
LAUDL002847 LAUDL002848 LAUDL002850
Calibration thermostats, ECO
Termostatos con circulación Sistemas de circulación abierta Circuladores de calibración
Calibration thermostatic bath with latest microprocessor technology and integrated cooling system. Working temperature range: −25 to 200 °C
  • Safety class III for operation with flammable and non-flammable liquids. Over-temperature cut-out adjustable via menu
  • Various pump with six adjustable performance levels
  • Easy control of the flow rate between internal and external circulation during operation without contact with the bath
  • USB interface as standard
  • 2-chamber construction for constant level in the thermosetting chamber
  • Vertical adjustment of the overflow