Stopcocks for burette, straight
Proveedor: LENZ, E. (Lenz Laborglas GmbH & CO.KG)
02700501
LENZ02700501EA 28.1 EUR
LENZ02700501
Stopcocks for burette, straight
Válvulas de cierre
NS stopcock made of borosilicate glass 3.3, facetted tip with plug of PTFE and screwthread retaining device.
  • Excellent chemical resistance
  • Excellent temperature resistance