Matraces Erlenmeyer con boca esmerilada estándar, ámbar
Proveedor: LENZ, E. (Lenz Laborglas GmbH & CO.KG)
03022022 03023028 03023037 03023046 03023049 03023052 03023058 03023070 03023073 81002951 302808 308606 81002066
LENZ03022022EA 18.5 EUR
LENZ03022022 201-0926 201-0927 201-0928 201-0929 201-0930 201-0931 201-0932 201-0933 LENZ81002951 LENZ302808 LENZ308606 LENZ81002066
Matraces Erlenmeyer con boca esmerilada estándar, ámbar
Matraces Matraces Erlenmeyer
DURAN® borosilicate glass, brown.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Coloured brown to protect light-sensitive substances. Volume graduations and labelling area.