Bridas, KF, con oliva de vidrio
Proveedor: LENZ, E. (Lenz Laborglas GmbH & CO.KG)
05106216 05106110 05106116 05107110 05107116 05107216
LENZ05106216EA 66.1 EUR
LENZ05106216 LENZ05106110 LENZ05106116 LENZ05107110 LENZ05107116 LENZ05107216
Bridas, KF, con oliva de vidrio
Adaptadores Adaptadores con juntas de rótula
Made of DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes